7-Bromo-1,3-benzoxazole 95%

Supplier: Ambeed
Warning

Synonyms: 7-Bromobenzo[d]oxazole

A189211-100MG A189211-250MG A189211-1G A189211-5G
103816-168EA 61 USD
103816-168 103846-762 76690-812 76690-814 77084-526 77084-528
7-Bromo-1,3-benzoxazole 95%
7-Bromo-1,3-benzoxazole

Precaución: for Research Use Only

Formula: C₇H₄BrNO
Boiling Pt: 251 °C (760 mm Hg)
Flash Pt: 106 °C
Storage Temperature: Freezer
MDL Number: MFCD08234619
CAS Number: 885270-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR