7-Bromobenzo[b]thiophene 96%
Supplier: Ambeed
|
Warning
|
A157319-100G
A157319-10G
A157319-250MG
A157319-25G
A157319-1G
A157319-5G
103804-986EA
17
USD
103804-986
103843-186
76783-676
76783-678
76783-680
77300-250
77319-004
77319-006
7-Bromobenzo[b]thiophene 96%
7-Bromobenzo[b]thiophene
Precaución: for Research Use Only
Formula:
C₈H₅BrS Boiling Pt: 284.7 °C (760 mm Hg) Flash Pt: 125.99 °C Storage Temperature: Ambient |
MDL Number:
MFCD06657729 CAS Number: 1423-61-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...