7-Bromo-6-chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine 97%

Supplier: Ambeed
Warning

A1002287-100MG A1002287-1G
103795-308EA 339 USD
103795-308 103821-020 76645-930 77282-460
7-Bromo-6-chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine 97%
7-Bromo-6-chloro-3,4-dihydro-2H-benzo[b][1,4]oxazine

Precaución: for Research Use Only

Formula: C₈H₇BrClNO
MW: 248.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21602798
CAS Number: 105679-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR