7-Bromo-5-fluoroquinoxaline 98%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-5-fluoro-1,4-benzodiazine

A314969-1G A314969-250MG
103825-052EA 198 USD
103825-052 76838-808 77015-200
7-Bromo-5-fluoroquinoxaline 98%
7-Bromo-5-fluoroquinoxaline

Precaución: for Research Use Only

Formula: C₈H₄BrFN₂
MW: 227.033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19053255
CAS Number: 1210048-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR