7-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-one 97%

Supplier: Ambeed
Warning

A302102-1G A302102-100MG A302102-250MG
103814-324EA 61 USD
103814-324 103835-480 76838-172 76988-776 76988-778
7-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-one 97%
7-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₆BrFO
MW: 229.05 g/mol
Boiling Pt: 315.6±42.0 °C (760 mm Hg)
Flash Pt: 144.7 °C
Storage Temperature: Ambient
MDL Number: MFCD16876114
CAS Number: 1260016-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR