7-Bromo-5-fluoroindole 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-5-fluoro-1H-indole

A149376-100MG A149376-250MG A149376-1G
103804-036EA 38 USD
103804-036 103842-154 76689-314 76689-316 77317-646
7-Bromo-5-fluoroindole 97%
7-Bromo-5-fluoroindole

Precaución: for Research Use Only

Formula: C₈H₅BrFN
MW: 214.04 g/mol
Boiling Pt: 315.1 °C (760 mm Hg)
Melting Pt: 20…25 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04037875
CAS Number: 408355-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR