7-Bromo-4-methoxyindole 95%

Supplier: Ambeed
Warning

A416265-100MG A416265-1G A416265-250MG
103827-310EA 247 USD
103827-310 76694-946 77307-178 77878-168
7-Bromo-4-methoxyindole 95%
7-Bromo-4-methoxyindole

Precaución: for Research Use Only

Formula: C₉H₈BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD04037872
CAS Number: 81224-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR