7-Bromoquinolin-4-ol 97%

Supplier: Ambeed
Danger

Synonyms: 7-Bromo-4-hydroxyquinoline

A275434-100MG A275434-10G A275434-250MG A275434-1G A275434-100G
103813-250EA 13 USD
103813-250 103853-176 76816-302 76816-304 76816-306 77093-028 77883-758
7-Bromoquinolin-4-ol 97%
7-Bromoquinolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Boiling Pt: 370.732 °C (760 mm Hg)
Melting Pt: 279…281 °C
Flash Pt: 178.013 °C
Storage Temperature: Ambient
MDL Number: MFCD06659037
CAS Number: 82121-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR