7-Bromo-4-chlorothieno[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A501577-1G A501577-10G A501577-250MG
103828-298EA 16 USD
103828-298 103839-386 76843-354 77169-330 77169-332
7-Bromo-4-chlorothieno[3,2-d]pyrimidine 97%
7-Bromo-4-chlorothieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₂BrClN₂S
MW: 249.5200 g/mol
Boiling Pt: 345.5 °C (760 mm Hg)
Melting Pt: 182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08273917
CAS Number: 31169-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR