7-Bromo-4-chloroquinoline 97%
Supplier: Ambeed
|
Warning
|
A126889-100MG
A126889-10G
A126889-250MG
A126889-1G
A126889-25G
A126889-5G
103800-868EA
8
USD
103800-868
77108-930
77108-932
77108-934
77313-106
77313-108
77313-110
7-Bromo-4-chloroquinoline 97%
7-Bromo-4-chloroquinoline
Precaución: for Research Use Only
Formula:
C₉H₅BrClN Melting Pt: 109…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684221 CAS Number: 75090-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...