7-Bromo-4-chloro-2-methylquinazoline 97%
Supplier: Ambeed
|
Warning
|
A178359-100MG
A178359-10G
A178359-250MG
A178359-1G
A178359-5G
103807-326EA
80
USD
103807-326
103845-666
76690-468
76690-470
76690-472
77083-088
77083-090
7-Bromo-4-chloro-2-methylquinazoline 97%
7-Bromo-4-chloro-2-methylquinazoline
Precaución: for Research Use Only
Formula:
C₉H₆BrClN₂ MW: 257.51 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09746300 CAS Number: 403850-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...