7-Bromo-4-azaindole 95%

Supplier: Ambeed
Warning

A108858-100MG A108858-1G A108858-250MG
103797-394EA 171 USD
103797-394 103823-184 77025-644 77310-010 77570-508
7-Bromo-4-azaindole 95%
7-Bromo-4-azaindole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD12024305
CAS Number: 1190318-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR