7-Bromo-4,4-dimethyl-1H-benzo[d][1,3]oxazin-2(4H)-one 98%

Supplier: Ambeed
Warning

A150070-100MG A150070-250MG A150070-5G A150070-1G
103804-114EA 661 USD
103804-114 77028-080 77028-082 77028-084 77299-770
7-Bromo-4,4-dimethyl-1H-benzo[d][1,3]oxazin-2(4H)-one 98%
7-Bromo-4,4-dimethyl-1H-benzo[d][1,3]oxazin-2(4H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO₂
Boiling Pt: 258 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD12755798
CAS Number: 1245643-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR