7-Bromo-3H-imidazo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

A133263-100MG A133263-250MG
103801-850EA 95 USD
103801-850 77027-174 77027-176
7-Bromo-3H-imidazo[4,5-b]pyridine 95%
7-Bromo-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846335
CAS Number: 1207174-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR