7-Bromo-1-tetralone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Bromo-3,4-dihydronaphthalen-1(2H)-one
A104347-100G
A104347-25G
A104347-500G
A104347-1G
A104347-5G
A104347-10G
103796-694EA
6
USD
103796-694
103822-406
76796-412
76796-414
76796-416
77309-274
77309-276
77592-106
7-Bromo-1-tetralone 98%
7-Bromo-1-tetralone
Precaución: for Research Use Only
Formula:
C₁₀H₉BrO Boiling Pt: 311.9 °C (760 mm Hg) Melting Pt: 78 °C Flash Pt: 103.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179287 CAS Number: 32281-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...