7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride 97%

Supplier: Ambeed
Warning

A379336-250MG A379336-1G
103826-644EA 28 USD
103826-644 103837-678 76817-646 76840-988
7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride 97%
7-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉BrClNO
MW: 250.5200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20278295
CAS Number: 1350738-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR