7-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyridine 98%

Supplier: Ambeed
Warning

A206300-250MG A206300-100MG
103817-702EA 58 USD
103817-702 103848-372 76825-754 77885-492
7-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyridine 98%
7-Bromo-2-methyl-[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
Storage Temperature: Ambient
MDL Number: MFCD12400851
CAS Number: 1159812-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR