7-Bromo-2-methoxyquinoxaline 95%

Supplier: Ambeed
Warning

A134775-100MG A134775-250MG
103802-046EA 79 USD
103802-046 77246-694 77246-696
7-Bromo-2-methoxyquinoxaline 95%
7-Bromo-2-methoxyquinoxaline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O
MW: 239.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03067025
CAS Number: 212327-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR