7-Bromo-2H-benzo[b][1,4]oxazin-3(4H)-one 95%

Supplier: Ambeed
Warning

A281836-10G A281836-250MG A281836-1G A281836-25G A281836-5G
103813-574EA 15 USD
103813-574 103853-526 76787-414 76787-416 77305-416 77305-418 77305-420
7-Bromo-2H-benzo[b][1,4]oxazin-3(4H)-one 95%
7-Bromo-2H-benzo[b][1,4]oxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₆BrNO₂
Flash Pt: 184.507 °C
Storage Temperature: Ambient
MDL Number: MFCD07774199
CAS Number: 321436-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR