7-Bromo-2-chloroquinoline 97%

Supplier: Ambeed
Warning

A190144-1G A190144-5G A190144-100MG A190144-10G A190144-250MG A190144-25G
103816-264EA 18 USD
103816-264 103846-862 77084-650 77084-652 77151-860 77151-862 77151-864 77302-212
7-Bromo-2-chloroquinoline 97%
7-Bromo-2-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
Melting Pt: 129…130 °C
Flash Pt: 150.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06738671
CAS Number: 99455-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR