7-Bromo-2,3-dihydro-1H-isoindol-1-one 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-2,3-dihydro-isoindol-1-one

A118784-250MG A118784-1G A118784-5G
103799-106EA 29 USD
103799-106 76971-374 77311-778 77311-780
7-Bromo-2,3-dihydro-1H-isoindol-1-one 97%
7-Bromo-2,3-dihydro-1H-isoindol-1-one

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MW: 212.043 g/mol
Boiling Pt: 452.2 °C (760 mm Hg)
Flash Pt: 227.3 °C
Storage Temperature: Ambient
MDL Number: MFCD11848600
CAS Number: 200049-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR