7-Bromo-1-naphthoic acid 95%

Supplier: Ambeed
Warning

A182908-100MG A182908-10G A182908-250MG A182908-1G
103815-584EA 139 USD
103815-584 103846-146 76985-798 76985-800 76985-802 77083-724
7-Bromo-1-naphthoic acid 95%
7-Bromo-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇BrO₂
MW: 251.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11502678
CAS Number: 51934-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR