7-Bromoindan-1-one 98%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-1-indanone, 7-Bromo-2,3-dihydro-1H-inden-1-one, 7-Bromo-2,3-dihydro-1-oxo-1H-indene

A184327-100G A184327-100MG A184327-250MG A184327-25G A184327-1G A184327-5G A184327-10G
103815-744EA 80 USD
103815-744 103846-306 77111-512 77111-514 77111-516 77111-518 77301-898 77301-900 77567-686
7-Bromoindan-1-one 98%
7-Bromoindan-1-one

Precaución: For research use only.

Formula: C₉H₇BrO
MW: 211.055 g/mol
Boiling Pt: 306.5 °C (760 mm Hg)
Flash Pt: 122.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06796574
CAS Number: 125114-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR