7-Bromo-1H-pyrrolo[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

A192248-10G A192248-250MG A192248-1G A192248-5G
103816-430EA 55 USD
103816-430 103847-036 76784-984 76784-986 77302-286 77302-288
7-Bromo-1H-pyrrolo[3,2-c]pyridine 95%
7-Bromo-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.0320 g/mol
Boiling Pt: 338 °C (760 mm Hg)
Flash Pt: 158 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781153
CAS Number: 902837-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR