7-Bromo-1H-indole-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A166214-10G
A166214-250MG
A166214-1G
A166214-5G
103806-028EA
25
USD
103806-028
103844-288
76690-046
76690-048
77320-532
77320-534
7-Bromo-1H-indole-2-carboxylic acid 98%
7-Bromo-1H-indole-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₆BrNO₂ MW: 240.0500 g/mol Boiling Pt: 470.9 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00744954 CAS Number: 16732-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...