7-Bromo-1H-indazole 97%

Supplier: Ambeed
Warning

A125177-100G A125177-10G A125177-250MG A125177-25G A125177-1G A125177-5G
103800-640EA 20 USD
103800-640 77161-558 77161-560 77161-562 77161-564 77312-784 77312-786
7-Bromo-1H-indazole 97%
7-Bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
Melting Pt: 126…130 °C
Flash Pt: 155.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07371564
CAS Number: 53857-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR