7-Bromo-1-heptene 98%

Supplier: Ambeed
Danger

A483724-10G A483724-1G A483724-25G A483724-5G
103828-158EA 6 USD
103828-158 103839-232 77116-436 77307-554 77307-556 77307-558
7-Bromo-1-heptene 98%
7-Bromo-1-heptene

Precaución: for Research Use Only

Formula: C₇H₁₃Br
MW: 177.085 g/mol
Boiling Pt: 178.7 °C (760 mm Hg)
Melting Pt: 0 °C
Flash Pt: 58.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631137
CAS Number: 4117-09-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR