4-Bromoisatoic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione

A222910-100MG A222910-10G A222910-250MG A222910-1G A222910-5G
103819-008EA 14 USD
103819-008 103849-886 76786-058 76786-060 76786-062 77088-496 77088-498
4-Bromoisatoic anhydride 97%
4-Bromoisatoic anhydride

Precaución: for Research Use Only

Formula: C₈H₄BrNO₃
MW: 242.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11113115
CAS Number: 76561-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR