7-Bromo-1,2-dihydroisoquinolin-3(4H)-one 95%

Supplier: Ambeed
Warning

A152617-250MG A152617-1G A152617-5G
103804-410EA 62 USD
103804-410 103842-582 77028-222 77318-204 77318-206
7-Bromo-1,2-dihydroisoquinolin-3(4H)-one 95%
7-Bromo-1,2-dihydroisoquinolin-3(4H)-one

Precaución: for Research Use Only

Formula: C₉H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD13193419
CAS Number: 943751-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR