7-Bromo-1,2,3,4-tetrahydroquinoline 95%

Supplier: Ambeed
Warning

A112828-100MG A112828-250MG A112828-1G A112828-5G
103798-116EA 43 USD
103798-116 103823-952 76810-004 76810-006 77310-776 77310-778
7-Bromo-1,2,3,4-tetrahydroquinoline 95%
7-Bromo-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀BrN
Storage Temperature: Refrigerator
MDL Number: MFCD08544270
CAS Number: 114744-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR