7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%

Supplier: Ambeed
Warning

A413917-1G A413917-25G A413917-5G A413917-100G A413917-10G
103827-266EA 12 USD
103827-266 103838-310 76841-870 76841-872 76841-874 77115-858 77115-860
7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%
7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClN
Flash Pt: 124.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094742
CAS Number: 220247-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR