7-Bromo-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: Ambeed
Warning

A139660-250MG A139660-1G
103802-760EA 63 USD
103802-760 77010-322 77315-856
7-Bromo-1,2,3,4-tetrahydroisoquinoline 97%
7-Bromo-1,2,3,4-tetrahydroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₁₀BrN
Boiling Pt: 282.908 °C (760 mm Hg)
Melting Pt: 32…35 °C
Flash Pt: 124.899 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739047
CAS Number: 17680-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR