7-Bromo[1,2,4]triazolo[4,3-a]pyridine 98%

Supplier: Ambeed
Warning

A101580-100MG A101580-25G A101580-5G A101580-250MG
103796-036EA 36 USD
103796-036 103821-722 76970-174 76970-176 77282-762 77592-072
7-Bromo[1,2,4]triazolo[4,3-a]pyridine 98%
7-Bromo[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
Storage Temperature: Ambient
MDL Number: MFCD09994418
CAS Number: 832735-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR