7-Bromo[1,2,4]triazolo[1,5-a]pyridine 97%

Supplier: Ambeed
Warning

A233350-10G A233350-250MG A233350-5G A233350-100MG
103810-886EA 42 USD
103810-886 103850-646 77031-112 77031-114 77089-604 77872-181
7-Bromo[1,2,4]triazolo[1,5-a]pyridine 97%
7-Bromo[1,2,4]triazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
Melting Pt: 108…108.5 °C
Storage Temperature: Ambient
MDL Number: MFCD10699200
CAS Number: 1053655-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR