7-Amino-5-fluoroindolin-2-one 95%
Supplier: Ambeed
|
Warning
|
A184594-100MG
A184594-250MG
A184594-5G
103815-762EA
94
USD
103815-762
103846-326
76872-866
76872-868
77083-944
7-Amino-5-fluoroindolin-2-one 95%
7-Amino-5-fluoroindolin-2-one
Precaución: for Research Use Only
Formula:
C₈H₇FN₂O MW: 166.152 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13193359 CAS Number: 945381-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...