7-Amino-4-methylcoumarin 98%

Supplier: Ambeed
Warning

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

A371201-100G A371201-10G A371201-10MG A371201-500G A371201-50MG A371201-25G A371201-5G A371201-1G
103826-476EA 8 USD
103826-476 103837-496 76803-592 76803-594 76803-596 76803-598 76803-600 76840-744 76840-746 77306-828
7-Amino-4-methylcoumarin 98%
7-Amino-4-methylcoumarin

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 378.3 °C (760 mm Hg)
Melting Pt: 223…226 °C
Flash Pt: 216.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006868
CAS Number: 26093-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR