7-Amino-3,4-dihydro-2H-isoquinolin-1-one 95%

Supplier: Ambeed
Warning

Synonyms: 7-Amino-3,4-dihydroisoquinolin-1(2H)-one

A136850-100MG A136850-250MG A136850-1G A136850-5G
103802-362EA 102 USD
103802-362 77162-132 77162-134 77315-280 77315-282
7-Amino-3,4-dihydro-2H-isoquinolin-1-one 95%
7-Amino-3,4-dihydro-2H-isoquinolin-1-one

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MW: 162.189 g/mol
Flash Pt: 253.7 °C
Storage Temperature: Ambient
MDL Number: MFCD08437645
CAS Number: 66491-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR