7-Amino-3,4-dihydro-2(1H)-quinolinone 97%

Supplier: Ambeed
Warning

A121045-10G A121045-250MG A121045-1G A121045-5G
103800-026EA 19 USD
103800-026 76688-118 76688-120 77297-810 77297-812
7-Amino-3,4-dihydro-2(1H)-quinolinone 97%
7-Amino-3,4-dihydro-2(1H)-quinolinone

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O
MW: 162.1885 g/mol
Boiling Pt: 403.1 °C (760 mm Hg)
Flash Pt: 197.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06809628
CAS Number: 22246-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR