7-Amino-2,3-dihydroinden-1-one 97%

Supplier: Ambeed
Warning

A495113-1G A495113-100MG A495113-250MG
103828-248EA 63 USD
103828-248 103839-328 76843-296 77143-984 77143-986
7-Amino-2,3-dihydroinden-1-one 97%
7-Amino-2,3-dihydroinden-1-one

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.17 g/mol
Boiling Pt: 325.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11035916
CAS Number: 628732-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR