7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one 95%

Supplier: Ambeed
Warning

A128088-10G A128088-25G A128088-5G A128088-1G A128088-250MG
103801-066EA 7 USD
103801-066 77026-922 77026-924 77298-262 77313-352 77871-587
7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one 95%
7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₃
Boiling Pt: 455.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02091565
CAS Number: 73942-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR