7,8-Dihydro-1,6-naphthyridin-5(6H)-one 95%

Supplier: Ambeed
Warning

A123203-100MG A123203-250MG
103800-362EA 398 USD
103800-362 77026-592 77026-594
7,8-Dihydro-1,6-naphthyridin-5(6H)-one 95%
7,8-Dihydro-1,6-naphthyridin-5(6H)-one

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Boiling Pt: 413.9 °C (760 mm Hg)
Melting Pt: 163…166 °C
Flash Pt: 204.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00928591
CAS Number: 155058-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR