7-(Trifluoromethyl)quinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A512878-100MG A512878-1G A512878-5G A512878-250MG
103828-424EA 135 USD
103828-424 103839-522 76695-800 76843-510 76843-512 77627-016
7-(Trifluoromethyl)quinoline-3-carboxylic acid 97%
7-(Trifluoromethyl)quinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₆F₃NO₂
MW: 241.1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042923
CAS Number: 71082-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR