7-(Trifluoromethyl)isatin 97%

Supplier: Ambeed
Warning

A246178-1G A246178-25G A246178-5G A246178-100G A246178-10G
103811-692EA 10 USD
103811-692 103851-520 77090-802 77090-804 77090-806 77261-906 77261-908
7-(Trifluoromethyl)isatin 97%
7-(Trifluoromethyl)isatin

Precaución: for Research Use Only

Formula: C₉H₄F₃NO₂
MW: 215.1300 g/mol
Melting Pt: 192…193 °C
Storage Temperature: Ambient
MDL Number: MFCD01248408
CAS Number: 391-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR