7-(Trifluoromethyl)-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A404354-100MG A404354-250MG A404354-5G A404354-1G
103827-108EA 265 USD
103827-108 77003-954 77003-956 77003-958 77307-088
7-(Trifluoromethyl)-1H-indole-2-carboxylic acid 95%
7-(Trifluoromethyl)-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD02664424
CAS Number: 172216-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR