7-(tert-Butoxy)-7-oxoheptanoic acid 98%
Supplier: Ambeed
|
Warning
|
A430437-10G
A430437-250MG
103827-510EA
107
USD
103827-510
103838-574
76648-928
76648-930
7-(tert-Butoxy)-7-oxoheptanoic acid 98%
7-(tert-Butoxy)-7-oxoheptanoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₂₀O₄ MW: 216.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30720967 CAS Number: 1469894-57-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...