7-(Bromomethyl)pentadecane 97%

Supplier: Ambeed
Warning

A271841-25G A271841-5G A271841-1000G A271841-100G A271841-10G A271841-500G A271841-1G
103813-070EA 9 USD
103813-070 103852-982 77092-770 77092-772 77238-756 77238-758 77238-760 77238-762 77305-100
7-(Bromomethyl)pentadecane 97%
7-(Bromomethyl)pentadecane

Precaución: for Research Use Only

Formula: C₁₆H₃₃Br
MW: 305.34 g/mol
Boiling Pt: 341.1 °C (760 mm Hg)
Flash Pt: 107.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00136231
CAS Number: 52997-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR