7-(Benzyloxy)-2,4-dichloro-6-methoxyquinazoline 95%

Supplier: Ambeed
Warning

A386428-100MG A386428-250MG
103826-798EA 318 USD
103826-798 103837-838 76990-064 76990-066
7-(Benzyloxy)-2,4-dichloro-6-methoxyquinazoline 95%
7-(Benzyloxy)-2,4-dichloro-6-methoxyquinazoline

Precaución: for Research Use Only

Formula: C₁₆H₁₂Cl₂N₂O₂
MW: 335.19 g/mol
Boiling Pt: 447.6±45.0 °C (760 mm Hg)
Flash Pt: 224.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16987836
CAS Number: 60771-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR