7-(Benzyloxy)-1H-indole-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A205491-1G A205491-5G A205491-100MG A205491-250MG
103817-634EA 68 USD
103817-634 103848-300 77086-516 77086-518 77097-534 77097-536
7-(Benzyloxy)-1H-indole-3-carboxylic acid 97%
7-(Benzyloxy)-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₃
MW: 267.28 g/mol
Boiling Pt: 531.1 °C (760 mm Hg)
Flash Pt: 275 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07369965
CAS Number: 24370-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR