7-Indoleboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
A217937-10G
A217937-250MG
A217937-1G
A217937-5G
103818-674EA
20
USD
103818-674
103849-406
76691-696
76691-698
77088-052
77088-054
7-Indoleboronic acid pinacol ester 97%
7-Indoleboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BNO₂ Boiling Pt: 396 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08669591 CAS Number: 642494-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...