6-Thioxo-6,9-dihydro-1H-purin-2(3H)-one 95%

Supplier: Ambeed
Warning

A285350-1G A285350-250MG
103813-670EA 84 USD
103813-670 103834-804 76837-338 77014-658
6-Thioxo-6,9-dihydro-1H-purin-2(3H)-one 95%
6-Thioxo-6,9-dihydro-1H-purin-2(3H)-one

Precaución: for Research Use Only

Formula: C₅H₄N₄OS
MW: 168.1765 g/mol
Melting Pt: 300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00143509
CAS Number: 2002-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR